Learn More
anti-RASGRP3, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP131501
Catalog No. NBP131501
Add to cart
Description
Description
RASGRP3 Polyclonal antibody specifically detects RASGRP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
RASGRP3
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Q8IV61
Calcium and DAG-regulated guanine nucleotide exchange factor III, GRP3KIAA0846ras guanyl-releasing protein 3, Guanine nucleotide exchange factor for Rap1, RAS guanyl releasing protein 3 (calcium and DAG-regulated)
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
25780
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Affinity Purified
RASGRP3
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human RASGRP3. The exact sequence is proprietary.
78 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Chicken (85%) and Rhesus Monkey (100%). Not yet tested in any other species.