Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASGRP3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528091

Catalog No. PIPA528091


Add to cart

Description

Description

PA5-28091 targets RASGRP3 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-28091 immunogen is synthetic peptide corresponding to a region within amino acids 1 and 44 of Human RASGRP3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Members of the RAS (see HRAS; MIM 190020) subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RASGRP3, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation (Rebhun et al., 2000 [PubMed 10934204]).
Specifications

Specifications

RASGRP3
Polyclonal
Unconjugated
RASGRP3
Liquid
RASGRP3
Synthetic peptide corresponding to a region within amino acids 1 and 44 of Human RASGRP3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.64 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q8IV61
GRP3, calcium and DAG-regulated guanine nucleotide exchange factor III, guanine nucleotide exchange factor for Rap1, ras guanyl-releasing protein 3
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
25780
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit