Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-2B4/CD244/SLAMF4, DyLight 350, Clone: 244F4, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP200223UV
Description
2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
2B4/CD244/SLAMF4 | |
Monoclonal | |
DyLight 350 | |
CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Mouse | |
IgG2a |
Western Blot, Flow Cytometry | |
244F4 | |
Western Blot, Flow Cytometry | |
CD244 | |
The immunogen for this antibody is CD244. | |
0.1 mL | |
Phospho Specific | |
51744 | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only