Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti- Activin RIB/ALK-4, Clone: 8J28, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236294

Catalog No. NBP236294

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Activin RIB/ALK-4
8J28
Unconjugated
PBS with No Preservative
activin A receptor, type IB, Activin receptor type IB, Activin receptor-like kinase 4, ACTRIB, ACVRLK4ACTR-IB, ALK-4, ALK4activin A receptor, type II-like kinase 4, EC 2.7.11, EC 2.7.11.30, Serine/threonine-protein kinase receptor R2, SKR2activin receptor type-1B
Rat
IgG2
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Western Blot
LYOPH
Western Blot 1:100 - 1:2000
Purified
Acvr1b
recombinant mouse ALK4 extracellular domain
Protein G purified
RUO
Primary
91
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only