Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti- Artemin, Clone: 6D32, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236297

Catalog No. NBP236297


Add to cart

Description

Description

Artemin Monoclonal antibody specifically detects Artemin in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Artemin
6D32
Unconjugated
PBS with No Preservative
artemin, ENOVIN, EVNneurotrophic factor, NBN, Neublastin
Rat
IgG2
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Immunohistochemistry, Western Blot
LYOPH
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Purified
ARTN
recombinant mouse Artemin
Protein G purified
RUO
Primary
9048
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only