Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-B7-1/CD80, Biotin, Clone: 1G10, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP128028
Description
B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, In vitro assay.Specifications
B7-1/CD80 | |
1G10 | |
Biotin | |
Purified | |
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
CD80 | |
Activated 5C2 cells | |
Monoclonal | |
Protein A or G purified | |
RUO | |
Mouse |
Flow Cytometry,Immunoprecipitation,In vitro assay | |
0.5mg/mL | |
Flow Cytometry <= 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500, In vitro assay | |
PBS with 0.05% Sodium Azide | |
941 | |
Rat | |
IgG2a Kappa | |
Primary | |
0.25 mg | |
Store at 4C. Do not freeze. | |
Mouse CD80 (B7-1), Mr 45-47 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only