Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti- CCL20/MIP-3 alpha, Clone: 11J22, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236316

Catalog No. NBP236316

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CCL20/MIP-3 alpha
11J22
Unconjugated
PBS with No Preservative
beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38
Rat
IgG2
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Blocking Assay, Western Blot
LYOPH
Western Blot 1:100 - 1:2000, Blocking/Neutralizing 50 - 200 molar excess
Purified
CCL20
recombinant mouse CCL-20
Protein G purified
RUO
Primary
6364
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only