Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Antibody (1D3), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP224965UV
Description
CD19 Monoclonal specifically detects CD19 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Flow (Cell Surface).Specifications
CD19 | |
Monoclonal | |
DyLight 350 | |
P25918 | |
CD19 | |
Mouse CD19 Transfected Cell Line | |
0.1 mL | |
Adaptive Immunity, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
930 | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
1D3 | |
Flow Cytometry, Flow (Cell Surface) | |
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
This antibody is reactive to Human. | |
Store at 4C in the dark. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction