Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1d Mouse anti-Rat, PE, Clone: WTH1 (also known as WTH-1), BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564632

Catalog No. BDB564632


Add to cart

Description

Description

The WTH1 monoclonal antibody specifically binds to rat CD1d and crossreacts with mouse CD1d (with stronger reactivity against mouse CD1d1 than CD1d2). CD1d is a type I transmembrane glycoprotein that is noncovalently associated with β2-microglobulin. CD1d is expressed on various cell types including subsets of thymocytes, T cells, B cells, monocytes, macrophages, dendritic cells, endothelial cells and epithelial cells. Although structurally similar to MHC Class I antigens, CD1d molecules are rather non-polymorphic and serve to present non-peptide antigens such as endogenous or microbial glycolipids to T lymphocytes (NKT cells). The WTH2 monoclonal antibody reportedly binds to a non-overlapping epitope on rat CD1d and also crossreacts with mouse CD1d when compared with WTH1. Both WTH1 and WTH2 reportedly interfere with antigen recognition by CD1d-restricted T cells.

Specifications

Specifications

CD1d
WTH1 (also known as WTH-1)
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG2a, κ
0.1mg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Cd1d; Cd1d1; Antigen-presenting glycoprotein CD1d
Affinity Purified
Rat CD1d Transfected Cell Line
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit