Learn More
anti-CD2, FITC, Clone: 12-15, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP126667
Catalog No. NBP126667
Add to cart
Description
Description
CD2 Monoclonal antibody specifically detects CD2 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, In vitro assay.
Specifications
Specifications
CD2
12-15
FITC
PBS with 0.05% Sodium Azide
CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5
Rat
IgG1 Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
Mouse CD2/LFA-2 (Mr 45-58 kDa)
Flow Cytometry, Immunoprecipitation, In vitro assay
0.5mg/mL
Flow Cytometry, Immunoprecipitation 1:10-1:500, In vitro assay
Purified
CD2
Crude plasma membranes of ESb cells
Protein A or G purified
RUO
Primary
914
Mouse