Learn More
anti-CD23/Fc epsilon RII, Alexa Fluor 700, Clone: 2G8, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP127907AF700
Catalog No. NBP127907Z
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD23/Fc epsilon RII
2G8
Flow Cytometry, ELISA, Immunohistochemistry-Frozen
Purified
FCER2
Fc-epsilon R isolated from B cell lymphoma cell line M12.4.5
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
CD23 low affinity IgE Fc receptor
ELISA, Flow Cytometry, Immunohistochemistry (Frozen)
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Rat
IgG2a Kappa
0.25 ml
Adaptive Immunity, Cancer, Immunology, Phospho Specific
Primary
2208
Mouse