Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD23/Fc epsilon RII, APC, Clone: 2G8, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP127907APC

Catalog No. NBP127907J

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CD23/Fc epsilon RII
2G8
Flow Cytometry
Purified
FCER2
Fc-epsilon R isolated from B cell lymphoma cell line M12.4.5
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
CD23 low affinity IgE Fc receptor
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Rat
IgG2a Kappa
0.25 ml
Adaptive Immunity, Cancer, Immunology, Phospho Specific
Primary
2208
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only