Learn More
anti-CD23/Fc epsilon RII, Clone: 2G8, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP127907
Catalog No. NBP127907
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD23/Fc epsilon RII
2G8
Flow Cytometry <= 1 ug/10^6 cells, ELISA 1:100-1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
FCER2
Fc-epsilon R isolated from B cell lymphoma cell line M12.4.5
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
CD23 low affinity IgE Fc receptor
Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Frozen)
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Rat
IgG2a Kappa
0.25 mg
Adaptive Immunity, Cancer, Immunology
Primary
2208
Mouse