Learn More
anti-CD23/Fc epsilon RII, FITC, Clone: 2G8, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP127908
Catalog No. NBP127908
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD23/Fc epsilon RII
2G8
FITC
PBS with 0.05% Sodium Azide
BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Rat
IgG2a Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
CD23 low affinity IgE Fc receptor
Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen)
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
FCER2
Fc-epsilon R isolated from B cell lymphoma cell line M12.4.5
Protein A or G purified
RUO
Primary
2208
Mouse