Learn More
anti-CD24, APC, Clone: 91, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP127916
Catalog No. NBP127916
Add to cart
Description
Description
CD24 Monoclonal antibody specifically detects CD24 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
Specifications
CD24
91
APC
PBS and a stabilizing agent with Sodium Azide
CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen
Rat
IgG2a Kappa
0.1 mg
Store at 4C. Do not freeze.
Monoclonal
Mouse heat stable antigen (HSA); Mr 35-45 kDa
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
0.1 mg/mL
Flow Cytometry <= 0.3 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
CD24
Mouse heat stable antigen (HSA); Mw 35-45 kDa
Protein A or G purified
RUO
Primary
100133941
Mouse