Learn More
anti-CD24, PE, Clone: 30-F1, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128130
Catalog No. NBP128130
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD24 Monoclonal antibody specifically detects CD24 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD24
30-F1
PE
PBS and a stabilizing agent with Sodium Azide
CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen
Rat
IgG2c
0.2 mg
Store at 4C in the dark.
Monoclonal
Mouse CD24/heat stable antigen (HSA)
Flow Cytometry
0.1 mg/mL
Flow Cytometry 0.2 ug/10^6 cells
Purified
CD24
Mouse thymus or spleen
Protein A or G purified
RUO
Primary
100133941
Mouse