Learn More
CD3 epsilon Antibody (C363.29B (YCD3-1)), Cy5, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP126690
Catalog No. NBP126690
Add to Cart
Description
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, In vitro assay.
Specifications
Specifications
CD3 epsilon
Monoclonal
0.1 mg/mL
Flow Cytometry < = 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, In vitro assay
CD3E
IL-4 producing Th2 cell lines including D10
0.1 mg
Innate Immunity
916
Mouse
IgG2b κ
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), In vitro Assay
C363.29B (YCD3-1)
Cyanine5
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Rat
Affinity Purified
RUO
Primary
Mouse CD3 epsilon (Mr 25 kD)
Store at 4C. Do not freeze.
Product Suggestions
Product Suggestions