Learn More
anti-CD38, PE, Clone: NIMR-5, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP127954
Catalog No. NBP127954
Add to cart
Description
Description
CD38 Monoclonal antibody specifically detects CD38 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
Specifications
CD38
NIMR-5
PE
PBS and a stabilizing agent with Sodium Azide
ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10
Rat
IgG2a Kappa
0.1 mg
Hematopoietic Stem Cell Markers, Stem Cell Markers
Primary
952
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
0.1 mg/mL
Flow Cytometry <= 0.2 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
CD38
BCL1 plasma membrane glycoproteins
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse