Learn More
CD4 Sheep anti-Rat, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF6439
Catalog No. AF6439
Add to cart
This item is not returnable. View return policy
Description
Description
CD4 Polyclonal antibody AF6439 Specifically detects CD4 in Rat samples. It is validated for Flow Cytometry, Western Blotting
Specifications
Specifications
CD4
LYOPH
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
P05540
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects rat CD4 in direct ELISAs and Western blots. In direct ELISAs, approximately 3% cross-reacitvity with recombinant feline CD4 is observed, and less than 1% cross-reactivity with recombinant human CD4 and recombinant canine CD4 is observed.
Rat
CyTOF, Flow Cytometry, Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CD4
Mouse myeloma cell line NS0-derived recombinant rat CD4 Lys28-Thr394 Accession # P05540
Immunogen affinity purified
RUO
Primary
920
Sterile PBS to a final concentration of 0.2 mg/mL.