Learn More
anti-CD44, Biotin, Clone: KM201, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP126582
Catalog No. NBP126582
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD44
KM201
Biotin
PBS with 0.05% Sodium Azide
Purified
CD44
(C57BL/6 x DBA/2)F1 mouse bone marrow-derived stromal clone BMS2
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
All isoforms of CD44/pgp-1 (Mr 80-95 kDa)
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.5mg/mL
Western Blot 1:100-1:2000, Flow Cytometry <= 2 ug/10^6 cells, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
P16070
CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I
Rat
IgG1
0.25 mg
Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
960
Mouse