Learn More
CD45 Rat anti-Mouse, Clone: I3/2.3, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP127970
Catalog No. NBP127970
Add to cart
Description
Description
CD45 Monoclonal antibody specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen).
Specifications
Specifications
CD45
I3/2.3
Unconjugated
BBS (pH 8.2) with No Preservative
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Rat
IgG2b
0.5 mg
Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
5788
Mouse
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
0.5 mg/mL
Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
PTPRC
T1M1 (Thy-1--c) cells
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse CD45 (T200 or leukocyte common antigen/LCA, all isoforms)