Learn More
CD45 Rat anti-Mouse, FITC, Clone: I3/2.3, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP127973
Catalog No. NBP127973
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD45 Monoclonal antibody specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen).
Specifications
Specifications
CD45
I3/2.3
FITC
PBS with Sodium Azide
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Rat
IgG2b
0.5 mg
Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
5788
Mouse
Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen)
0.5 mg/mL
Flow Cytometry < 1ug/1000000 cells, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
PTPRC
T1M1 (Thy-1--c) cells
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse CD45 (T200 or leukocyte common antigen/LCA, all isoforms)