Learn More
anti-CD45RB, Clone: C363.16A, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP127986
Catalog No. NBP127986
Add to cart
Description
Description
CD45RB Monoclonal antibody specifically detects CD45RB in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
CD45RB
C363.16A
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Rat
IgG2a Kappa
0.25 mg
Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
5788
Mouse
Flow Cytometry, Western Blot
0.5 mg/mL
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000
Purified
PTPRC
Cloned mouse Th cell line
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
An exon B-dependent epitope of CD45 glycoprotein