Learn More
anti-CD45RC, Clone: GL24, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP127990
Catalog No. NBP127990
Add to cart
Description
Description
CD45RC Monoclonal antibody specifically detects CD45RC in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation.
Specifications
Specifications
CD45RC
GL24
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Rat
IgM Kappa
0.5 mg
Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
5788
Mouse
Flow Cytometry, Immunoprecipitation
0.5 mg/mL
Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500
Purified
PTPRC
Activated DBA/2 B cells
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
An exon C-dependent epitope of CD45 glycoprotein