Learn More
anti-CD5, PE, Clone: 4H8E6, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP126706
Catalog No. NBP126706
Add to cart
Description
Description
CD5 Monoclonal antibody specifically detects CD5 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
Specifications
CD5
4H8E5
PE
PBS and a stabilizing agent with Sodium Azide
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
Rat
IgG2a Kappa
0.1 mg
Store at 4C. Do not freeze.
Monoclonal
CD5/Lyt-1, Mr 67 kDa
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
0.1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry <= 0.2 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
CD5
The immunogen is a peptide made to the amino acid region CD5
Protein A or G purified
RUO
Primary
921
Mouse