Learn More
anti-CD68/SR-D1, Alexa Fluor 405, Clone: FA-11, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233337AF405
Catalog No. NBP233337W
Add to cart
Description
Description
CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
CD68/SR-D1
FA-11
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
CD68
Purified ConA acceptor glycoproteins from the P815 cell line.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1
Rat
IgG2a
0.1 ml
Cell Biology, Cytokine Research, Immunology
Primary
968