Learn More
anti-CD90.2, Clone: 30-H12, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128036
Catalog No. NBP128036
Add to cart
Description
Description
CD90.2 Monoclonal antibody specifically detects CD90.2 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay, Immunocytochemistry.
Specifications
Specifications
CD90.2
30-H12
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Rat
IgG2b Kappa
0.5 mg
Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
7070
Mouse
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, In vitro assay
0.5 mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10 - 1:500, In vitro assay, Immunocytochemistry
Purified
THY1
Mouse thymus or spleen
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse CD90.2/Thy-1.2 alloantigen, Mr 18 kDa