Learn More
anti-CD90.2, FITC, Clone: 30-H12, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128037
Catalog No. NBP128037
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD90.2 Monoclonal antibody specifically detects CD90.2 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, In vitro assay, Immunocytochemistry.
Specifications
Specifications
CD90.2
30-H12
FITC
PBS with Sodium Azide
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Rat
IgG2b Kappa
0.5 mg
Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
7070
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), In vitro assay, Immunocytochemistry
0.1 mg/mL
Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay, Immunocytochemistry
Purified
THY1
Mouse thymus or spleen
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse CD90.2/Thy-1.2 alloantigen, Mr 18 kDa