Learn More
anti-CD90/Thy1, Biotin, Clone: G7, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128033
Catalog No. NBP128033
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Paraffin, In vitro assay.
Specifications
Specifications
CD90/Thy1
G7
Biotin
PBS with Sodium Azide
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Rat
IgG2c Kappa
0.5 mg
Hematopoietic Stem Cell Markers, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
7070
Mouse
Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), In vitro assay
0.5 mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, In vitro assay
Affinity Purified
THY1
Mouse T cell hybridoma C6/G8
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse CD90 (Thy-1.1 & Thy-1.2 alloantigens/Thy-1 epitope region A); Mr 28-32 kDa