Learn More
anti-CD94, Clone: 18D3, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128119
Catalog No. NBP128119
Add to cart
Description
Description
CD94 Monoclonal antibody specifically detects CD94 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD94
18D3
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor
Rat
IgG2a Kappa
0.5 mg
Innate Immunity
Primary
3824
Mouse
Flow Cytometry
0.5 mg/mL
Flow Cytometry 1 ug/10^6 cells
Purified
KLRD1
CHO cells transfected with the B6 allele of CD94
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse CD94 receptor subunit, Mr 70 kDa