Learn More
anti- Cystatin B/Stefin B, Clone: 45N2, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236033
Catalog No. NBP236033
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Cystatin B/Stefin B
45N2
Unconjugated
PBS with No Preservative
CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B
Rat
IgG2
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Immunohistochemistry, Western Blot
LYOPH
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Purified
CSTB
mouse recombinant Cystatin B
Protein G purified
RUO
Primary
1476
Mouse