Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F4/80 Antibody (CI-A3-1), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB600404APC
Description
F4/80 Monoclonal specifically detects F4/80 in Human, Mouse, C. elegans, E. coli samples. It is validated for Flow Cytometry.Specifications
F4/80 | |
Monoclonal | |
APC | |
ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 | |
Rat | |
160 kDa | |
0.125 mL | |
Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
2015 | |
Human, Mouse, C. elegans, E. coli | |
Purified |
Flow Cytometry | |
CI-A3-1 | |
Flow Cytometry | |
ADGRE1 | |
Thioglycollate stimulated peritoneal macrophages from C57BL/6 mice. | |
Protein G purified | |
RUO | |
Primary | |
This recognizes mouse F4/80, a 160kD glycoprotein expressed by murine macrophages. | |
Store at 4C in the dark. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction