Learn More
anti-ICAM-2/CD102, FITC, Clone: 3C4, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128218
Catalog No. NBP128218
Add to cart
Description
Description
ICAM-2/CD102 Monoclonal antibody specifically detects ICAM-2/CD102 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay, Immunocytochemistry.
Specifications
Specifications
ICAM-2/CD102
3C4
FITC
PBS with 0.05% Sodium Azide
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2
Rat
IgG2a Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
Mouse
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, In vitro assay
0.5mg/mL
Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay, Immunocytochemistry
Purified
ICAM2
COS cells transfected with mouse ICAM-2 cDNA and BW5147 AKR/J mouse thymoma cell line
Protein A or G purified
RUO
Primary
3384