Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-RAT IGG (H+L) AB BIOTIN  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52600042
Affinity purified polyclonal antibody to Rat IgG, heavy and light chains (whole IgG) made in Goat and biotinylated. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-70