Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-RAT IGG (H+L) AB DL488  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300387
Affinity purified polyclonal antibody to Rat IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 488. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-670-81