Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-RAT IGG(H+L) AB MSA  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52100180
Affinity purified polyclonal antibody to Rat IgG, both heavy chain and light chains (whole IgG) made in Goat. Product has been cross-adsorbed with mouse immunoglobulin to reduce cross-reactivity. Product is in lyophilized form.
Catalog No. 50-668-21