Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Kappa Light Chain Rat anti-Rat, Alexa Fluor 488, Clone: KLH/G1-2-2, Isotype Control, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP151817AF488

Catalog No. NBP151817X


Add to cart

Description

Description

IgG1 Kappa Light Chain Monoclonal antibody specifically detects IgG1 Kappa Light Chain in Rat samples. It is validated for Flow Cytometry, ELISA, Fluorophore-linked immunosorbent assay.
Specifications

Specifications

IgG1 Kappa Light Chain
KLH/G1-2-2
Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Purified
A bacterially expressed fusion protein containing amino acid residues 1-317 of mouse TRANCE was used as immunogen.
500 Tests
Store at 4C in the dark.
Primary
Rat
ELISA, Flow Cytometry, Immunofluorescence
Alexa Fluor 488
50 mM Sodium Borate. with 0.05% Sodium Azide
Mouse
IgG1 Kappa
RUO
Isotype Control
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only