Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rat IgG1, kappa L Chain, PerCP, Clone: KLH/G1-2-2 Isotype Control, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP151817PCP

Catalog No. NBP151817P


Add to cart

Description

Description

IgG1 Kappa Light Chain Monoclonal antibody specifically detects IgG1 Kappa Light Chain in Rat samples. It is validated for Flow Cytometry, ELISA, Fluorophore-linked immunosorbent assay.
Specifications

Specifications

IgG1 Kappa Light Chain
KLH/G1-2-2
Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Liquid
Keyhole limpet hemocyanin (KLH)
500 Tests
Store at 4C in the dark.
Primary
Immunoglobulin reacts with Keyhole limpet hemocyanin (KLH).
ELISA, Flow Cytometry, Immunofluorescence
PerCP
PBS. with 0.05% Sodium Azide
Rat
IgG1 Kappa
RUO
Isotype Control
Monoclonal
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only