Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rat IgG2a, Clone: KLH/G2a-1-1 Isotype Control, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP151104

Catalog No. NBP151104


Add to cart

Description

Description

IgG2a Monoclonal antibody specifically detects IgG2a in Rat samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Fluorophore-linked immunosorbent assay, Control, Block/Neutralize.
Specifications

Specifications

IgG2a
KLH/G2a-1-1
Unconjugated
PBS. with No Preservative
Rat
IgG2a
RUO
Isotype Control
Monoclonal
Blocking Assay, ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
0.5 mg/mL
Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, Control
Concentrated
KLH
0.25 mg
Store at -20C.
Primary
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only