Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IgG2b Kappa Light Chain Isotype Control (149/10H5), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143323UV
Description
IgG2b Kappa Light Chain Monoclonal specifically detects IgG2b Kappa Light Chain in Rat samples. It is validated for Flow Cytometry.Specifications
IgG2b Kappa Light Chain | |
Monoclonal | |
DyLight 350 | |
50 mM Sodium Borate. with 0.05% Sodium Azide | |
500 Tests | |
Isotype Control (Primary) | |
Store at 4C in the dark. | |
IgG2b κ |
Flow Cytometry | |
149/10H5 | |
Flow Cytometry 1:10 - 1:1000 | |
Rat | |
RUO | |
Rat | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction