Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rat IgM, Fluorescein, Isotype Control, Novus Biologicals™

Rat Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197202

Catalog No. NBP197202


Add to cart

Description

Description

IgM antibody specifically detects IgM in Rat samples. It is validated for SDS-Page.
Specifications

Specifications

IgM
Fluorescein
0.06 M Potassium Phosphate, 0.45 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
IgM
0.5 ml
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Rat Serum and anti-Rat IgM (mu chain specific). No reaction was observed against anti-Rat IgG F(c). Some light chain cross reactivity will occur with anti-Rat IgG.
Rat
SDS-Page
SDS-Page 1:100 - 1:2000
Lyophilized
Rat
Size Exclusion Chromatography
RUO
Isotype Control
3507
Reconstitute with 0.5 ml deionized water (or equivalent) to desired concentration.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only