Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rat IgM, kappa L Chain, DyLight 405, Clone: RGM Isotype Control, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143320V

Catalog No. NBP143320V


Add to cart

Description

Description

IgM Kappa Light Chain Monoclonal antibody specifically detects IgM Kappa Light Chain in Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

IgM Kappa Light Chain
RGM
Flow Cytometry 1:10 - 1:1000
Liquid
IgM Kappa
RUO
Isotype Control
Monoclonal
Flow Cytometry
DyLight 405
50 mM Sodium Borate. with 0.05% Sodium Azide
Rat
500 Tests
Store at 4C in the dark.
Primary
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only