Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IL-20 R beta/FNDC6, PerCP, Clone: 20RNTC, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP200466PCP
Description
IL-20 R beta/FNDC6 Monoclonal specifically detects IL-20 R beta/FNDC6 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
IL-20 R beta/FNDC6 | |
Monoclonal | |
PerCP | |
FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923 | |
Rat | |
Affinity Purified | |
RUO | |
53833 | |
Store at 4C in the dark. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
20RNTC | |
Flow Cytometry | |
IL20RB | |
The immunogen for this antibody is FNDC6 (IL-20R2). | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only