Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IL-6, Clone: MP5-20F3, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142355

Catalog No. NBP142355


Add to cart

Description

Description

IL-6 Monoclonal antibody specifically detects IL-6 in Mouse samples. It is validated for ELISA.
Specifications

Specifications

IL-6
MP5-20F3
ELISA 1:100-1:2000
Affinity Purified
IL6
COS-7-expressed, recombinant mouse IL-6
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse
ELISA
Unconjugated
PBS (pH 7.2) with 0.09% Sodium Azide
B cell stimulatory factor-2, B-cell differentiation factor, BSF-2, BSF2CTL differentiation factor, CDF, HGFHSFIFNB2Hybridoma growth factor, IFN-beta-2, IL-6B-cell stimulatory factor 2, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6
Rat
IgG1 Kappa
0.05 mg
Adaptive Immunity, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Primary
3569
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only