Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 4 beta 7/LPAM-1 Antibody (DATK32), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143283UV
Description
Integrin alpha 4 beta 7/LPAM-1 Monoclonal specifically detects Integrin alpha 4 beta 7/LPAM-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 4 beta 7/LPAM-1 | |
Monoclonal | |
DyLight 350 | |
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
DATK32 | |
Flow Cytometry | |
ITGA4 | |
The immunogen for this antibody was LPAM1. | |
0.1 mL | |
Innate Immunity | |
3676 | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction