Learn More
anti-Integrin alpha 4/CD49d, Biotin, Clone: PS/2, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP126663
Catalog No. NBP126663
Add to cart
Description
Description
Integrin alpha 4/CD49d Monoclonal antibody specifically detects Integrin alpha 4/CD49d in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Integrin alpha 4/CD49d
PS/2
Biotin
PBS with Sodium Azide
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Rat
IgG2b Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
CD49d, the alpha 4 chain (100 kDa) of the VLA-4 integrin heterodimer (Mr 150-kDa)
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
0.5mg/mL
Flow Cytometry <= 2 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
ITGA4
P815 cells
Protein A or G purified
RUO
Primary
3676
Human, Mouse