Learn More
anti-Integrin alpha L/CD11a, Clone: M17/4, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP141264
Catalog No. NBP141264
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Integrin alpha L/CD11a
M17/4
Unconjugated
PBS (pH 7.4) with 15mM Sodium Azide
CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1
Rat
IgG2a
Protein A purified
RUO
Store at 4C. Do not freeze.
Monoclonal
The rat monoclonal antibody M17/4 reacts with CD11a (alpha-subunit of murine LFA-1), a 180 kDa type I transmembrane glycoprotein expressed on B and T lymphocytes, monocytes, macrophages, neutrophils, basophils and eosinophils.
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay
1 mg/mL
Flow Cytometry 1 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:50, Immunohistochemistry-Frozen 1:200, Functional, Flow (Cell Surface), CyTOF-ready
Purified
ITGAL
C57BL/6 mouse splenic secondary cytotoxic T lymphocytes
180 kDa
0.1 mg
Adaptive Immunity, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers
Primary
3683
Mouse