Learn More
anti-Integrin alpha M/CD11b, FITC, Clone: M1/70, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP127882
Catalog No. NBP127882
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD11b
M1/70
FITC
PBS with 0.05% Sodium Azide
antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, CD11Bintegrin, alpha M (complement component receptor 3, alpha; also known as CD11b(p170), macrophage antigen alpha polypeptide), Cell surface glycoprotein MAC-1 subunit alpha, CR-3 alpha chain, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
Rat
IgG2b Kappa
0.25 mg
Innate Immunity
Primary
3684
Mouse
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay, Immunocytochemistry
Purified
ITGAM
C57BL/10 mouse splenic T cells and concanavalin A-activated C57BL/10 splenocytes
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
alpha M subunit (Mr 170 kDa) of the CD11b/CD18 heterodimer