Learn More
anti-Integrin beta 7, DyLight 755, Clone: 2G18, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11094147IR
Catalog No. NB11094147I
Add to cart
Description
Description
Integrin beta 7 Monoclonal specifically detects Integrin beta 7 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Integrin beta 7
2G18
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
ITGB7
Cell lysates of mouse lymphocytes
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
This was selected for its ability to detect mouse CD45 positive leukocytes in IHC.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7
Rat
IgG2
0.1 ml
Signal Transduction
Primary
3695
Mouse