Learn More
anti-KLRC1/2/3, DyLight 350, Clone: 20d5, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128100UV
Catalog No. NBP128100UV
Add to cart
Description
Description
KLRC1/2/3 Monoclonal antibody specifically detects KLRC1/2/3 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
KLRC1/2/3
20d5
Flow Cytometry
Purified
KLRC1
CHO cells transfected with B6 allele of NKG2A gene
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse NKG2 (Mr approx. 43 kDa)
Flow Cytometry
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B
Rat
IgG2a Kappa
0.25 ml
Signal Transduction
Primary
3821
Mouse